Copper wire, 1.5mm (0.06in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Copper wire, 1.5mm (0.06in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Copper wire, 1.5mm (0.06in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Copper wire, 1.5mm (0.06in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper wire, 1.5mm (0.06in) dia, 99.9% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF044549.G925 m
Catalog number ALF044549.G9
Price (MYR)
772.00
Add to cart
Quantity:
25 m
Request bulk or custom format
Price (MYR)
772.00
Add to cart
Specifications
Chemical Name or MaterialCopper wire
Name Note1.5mm (0.06 in.) dia.
CAS7440-50-8
Recommended StorageAmbient temperatures
EINECS Number231-159-6