Calcium D-gluconate gel, 2.5% w/w aq. soln., Thermo Scientific Chemicals
Calcium D-gluconate gel, 2.5% w/w aq. soln., Thermo Scientific Chemicals
Calcium D-gluconate gel, 2.5% w/w aq. soln., Thermo Scientific Chemicals
Calcium D-gluconate gel, 2.5% w/w aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium D-gluconate gel, 2.5% w/w aq. soln., Thermo Scientific Chemicals
CAS: 299-28-5 | C12H22CaO14 | 430.372 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
60 g
30 g
Catalog number ALF044542.TJ
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
60 g
Request bulk or custom format
Chemical Identifiers
CAS2414-98-4
IUPAC Namemagnesium(2+) diethanolate
Molecular FormulaC4H10MgO2
InChI KeyXDKQUSKHRIUJEO-UHFFFAOYSA-N
SMILES[Mg++].CC[O-].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl97.0 to 103.0 %
Appearance (Color)White to light grey
Infrared spectrumConforms
Appearance (Form)Powder
Calcium D-gluconate gel is used to treat hydrofluoric acid burns. It is used as a cardioprotective agent in hyperkalemia, antidote for black widow spider envenomation and an antidote for magnesium sulfate toxicity. It is also involved in the treatment of hypocalcemia. Further, it is used as skin conditioning as well as provides the source of calcium ions.