Nitric acid, 2.0N Standardized Solution, Thermo Scientific Chemicals
Nitric acid, 2.0N Standardized Solution, Thermo Scientific Chemicals
Nitric acid, 2.0N Standardized Solution, Thermo Scientific Chemicals
Nitric acid, 2.0N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitric acid, 2.0N Standardized Solution, Thermo Scientific Chemicals
CAS: 7697-37-2 | HNO3 | 63.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number ALF044528.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS128-08-5
IUPAC Name1-bromopyrrolidine-2,5-dione
Molecular FormulaC4H4BrNO2
InChI KeyPCLIMKBDDGJMGD-UHFFFAOYSA-N
SMILESBrN1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormPowder or crystalline powder
Assay (Iodometric Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Melting Point (clear melt)173.0-182.0?C
View more
Nitric acid, 2.0N Standardized Solution is used for volumetric analysis in laboratories.