n-Hexadecane, 99.5+%, Thermo Scientific Chemicals
n-Hexadecane, 99.5+%, Thermo Scientific Chemicals
n-Hexadecane, 99.5+%, Thermo Scientific Chemicals
n-Hexadecane, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexadecane, 99.5+%, Thermo Scientific Chemicals
CAS: 544-76-3 | C16H34 | 226.448 g/mol
Have Questions?
Catalog NumberQuantity
ALF044511.1425 g
Catalog number ALF044511.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS69045-79-0
IUPAC Name2-chloro-5-iodopyridine
Molecular FormulaC5H3ClIN
InChI KeyQWLGCWXSNYKKDO-UHFFFAOYSA-N
SMILESClC1=CC=C(I)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)95.0-101.0?C
Employed in industry as Adsorbents, Fuels and fuel additives. Also in Fabric, Textile, and Leather Products. Solvent, organic intermediate, ignition standard for diesel fuels. Fuels and Related Products. Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.