Fullerene, buckytube/nanotube, single walled, Thermo Scientific Chemicals
Fullerene, buckytube/nanotube, single walled, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fullerene, buckytube/nanotube, single walled, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
0.25 g
Catalog number ALF044508.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1459-93-4
IUPAC Name1,3-dimethyl benzene-1,3-dicarboxylate
Molecular FormulaC10H10O4
InChI KeyVNGOYPQMJFJDLV-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(=CC=C1)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormFused solid or flakes
Assay (GC)≥97.5%
Melting Point (clear melt)65.0-72.0?C
Carbon nanotubes enhance the electrical conductivity and the mechanical properties. It is also used in chemical research. It is suitable as a hydrogen storage material. It is an ideal super capacitor electrode materials, It is used in the preparation of super composites. carbon nanotubes have excellent field emission performance and used in heavy weight cathode tube technology. It is also applied to naanometer electronic, catalytic fiber machine. Nanotubes will have an important role in nanotechnology engineering. carbon nanotube also can be used in making molecule bearings and nano robot.