Phosphorus(III) chloride, 98% min, Thermo Scientific Chemicals
Phosphorus(III) chloride, 98% min, Thermo Scientific Chemicals
Phosphorus(III) chloride, 98% min, Thermo Scientific Chemicals
Phosphorus(III) chloride, 98% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorus(III) chloride, 98% min, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALF044436.18
Price (MYR)
97.42
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
97.42
Add to cart
Specifications
Boiling Point76°C
Chemical Name or MaterialPhosphorus(III) chloride
Melting Point°C to 111.8°C
CAS7719-12-2
ColorYellow
View more
Phosphorus(III) chloride is used as a reagent in organic synthesis. It is utilized in the conversion of primary alcohol and secondary alcohol to alkyl chlorides. It acts as a precursor to phosphorous pentachloride, phosphorous oxychloride, thiophosphoryl chloride and triphenylphosphine. It is also used to prepare phosphite esters, which is used Horner-Wadsworth-Emmons reactions.