Oxalic acid, anhydrous, 98%, Thermo Scientific Chemicals
Oxalic acid, anhydrous, 98%, Thermo Scientific Chemicals
Oxalic acid, anhydrous, 98%, Thermo Scientific Chemicals
Oxalic acid, anhydrous, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid, anhydrous, 98%, Thermo Scientific Chemicals
CAS: 144-62-7 | C2H2O4 | 90.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALF044410.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS5351-17-7
IUPAC Name4-aminobenzohydrazide
Molecular FormulaC7H9N3O
InChI KeyWPBZMCGPFHZRHJ-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point224°C to 228°C
HPLC>=94 %
Infrared spectrumConforms
Appearance (Color)White to beige
Appearance (Form)Powder or crystals
Oxalic acid is used as reducing agent for photographic films, as mordant in dyeing process, as bleaching agent for removal of rust through complex formation, and as grinding agent during polishing of marble. It is used in waste water treatment to remove calcium. The conjugate base of oxalic acid acts as a chelating agent for metal ions. It is used as a catalyst for esterifications in synthetic organic chemistry and as an analytical reagent.