Silica gel desiccant, -6+12 mesh granules, Thermo Scientific Chemicals
Silica gel desiccant, -6+12 mesh granules, Thermo Scientific Chemicals
Silica gel desiccant, -6+12 mesh granules, Thermo Scientific Chemicals
Silica gel desiccant, -6+12 mesh granules, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silica gel desiccant, -6+12 mesh granules, Thermo Scientific Chemicals
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number ALF044387.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilica gel desiccant
ColorWhite
Physical FormGranular
CAS7631-86-9
Recommended StorageAmbient temperatures
View more
Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.