Silica gel desiccant, -3+8 Mesh Granules, Thermo Scientific Chemicals
Silica gel desiccant, -3+8 Mesh Granules, Thermo Scientific Chemicals
Silica gel desiccant, -3+8 Mesh Granules, Thermo Scientific Chemicals
Silica gel desiccant, -3+8 Mesh Granules, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silica gel desiccant, -3+8 Mesh Granules, Thermo Scientific Chemicals
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number ALF044386.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS67627-18-3
IUPAC Nametripotassium (Z)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5',7-trisulfonate
Molecular FormulaC16H7K3N2O11S3
InChI KeyXOSMXDUITYWYGR-JRYLAINFSA-K
SMILES[K+].[K+].[K+].[O-]S(=O)(=O)C1=CC=C2NC(C(=O)C2=C1)=C1/NC2=C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)Dark blue
Molar extinction coefficient>= 15000 (597-603 nm)
Molar extinction coefficient>= 17000 (250-256 nm)
View more
Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.