Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals
CAS: 7440-18-8 | Ru | 101.07 g/mol
Have Questions?
Catalog NumberQuantity
ALF044338.22100 g
Catalog number ALF044338.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS15282-88-9
IUPAC Nameλ²-lead(2+) bis(2,4-dioxopentan-3-ide)
Molecular FormulaC10H14O4Pb
InChI KeyVJRBSSRKZYRKTG-UHFFFAOYSA-N
SMILES[Pb++].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to yellow to brown
FormSolid
Identification (FTIR)Conforms
Ruthenium, 5% on activated carbon powde is used in solar cells, which turn light energy into electrical energy.