Sodium metaperiodate, ACS, 99.8-100.3%, Thermo Scientific Chemicals
Sodium metaperiodate, ACS, 99.8-100.3%, Thermo Scientific Chemicals
Sodium metaperiodate, ACS, 99.8-100.3%, Thermo Scientific Chemicals
Sodium metaperiodate, ACS, 99.8-100.3%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium metaperiodate, ACS, 99.8-100.3%, Thermo Scientific Chemicals
Maximum level of impurities: Other halogens (as Cl) 0.02%, Mn 3ppm
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF044309.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS599-04-2
IUPAC Name3-hydroxy-4,4-dimethyloxolan-2-one
Molecular FormulaC6H10O3
InChI KeySERHXTVXHNVDKA-UHFFFAOYNA-N
SMILESCC1(C)COC(=O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)88-93?C
Optical Rotation-48? to -52? (c=2 in water)
Appearance (Color)White
FormCrystalline powder or crystals
Identification (FTIR)Conforms
View more
Sodium metaperiodate is used as an oxidizing reagent involved in the oxidation of cellulose. It is involved in the cleavage of vicinal diols to prepare two aldehydes. This oxidation is commonly used in selectively label RNA, saccharides and other compounds with vicinal diols.