Calcium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Calcium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Calcium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Calcium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 10043-52-4 | CaCl2 | 110.98 g/mol
Have Questions?
Catalog NumberQuantity
ALF044280.30250 g
Catalog number ALF044280.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS8007-47-4
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.5200-1.5310 @ 20°C
Appearance (Color)Clear colorless to yellow to green or pale pink to pale brown
FormViscous liquid
Calcium chloride is used as a drying and dehydrating agent for organic liquids and gases and for solids in desiccators. Common applications include brine for refrigeration plants and ice and dust control on roads. Use of calcium chloride and urea has been reported to significantly increase average fruit weight and ascorbic acid content in pomegranate. The strawberry fruit storability can also be improved by edible coating of calcium chloride. It is used in drying tubes in research laboratories, salt/chemical based dehumidifiers, fire-extinguishers, plastics, ceramic slipware, as flux in the Davy process for the manufacture of sodium and to increase water hardness in swimming pool. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. CaCl2-Pybox (pyridine flanked by two oxazoline groups) has been reported to be an efficient chiral catalyst for asymmetric 1,4-addition reactions of 1,3-dicarbonyl compounds with nitroalkenes, affording gamma-nitro carbonyl compounds in high enantioselectivities.