Zinc oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Zinc oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Zinc oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Zinc oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Zinc oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc oxide, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 1314-13-2 | OZn | 81.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Catalog number ALF044263.A7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Request bulk or custom format
Chemical Identifiers
CAS166953-64-6
IUPAC Namebenzyl 4-bromopiperidine-1-carboxylate
Molecular FormulaC13H16BrNO2
InChI KeyBFWKSPOSMPKTSV-UHFFFAOYSA-N
SMILESBrC1CCN(CC1)C(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to light brown
Appearance (Form)Liquid
Infrared spectrumConforms
HPLC>=95.0 %
NMR (1H-NMR)Consistent with structure
Zinc oxide is used as a sun screener. It serves as an additive in products such as cigarette filters, plastics, anticorrosive coatings, ceramics, glass, cement, lubricants, ointments, adhesives, sealants, pigments, batteries, ferrites, fire retardants, and first-aid tapes. It plays an important role in textiles as a UV-absorber. As a wide-bandgap semiconductor with favorable properties, it has the potential for use in transparent electrodes, in nanorod sensors, in liquid crystal displays, as thin-film transistors and light emitting diodes, and in heat-protecting windows.