Palladium nitrate, Matrix Modifier Solution, Specpure™
Palladium nitrate, Matrix Modifier Solution, Specpure™
Thermo Scientific Chemicals
Palladium nitrate, Matrix Modifier Solution, Specpure™
1% Pd in 10% HNO3
Have Questions?
Catalog NumberQuantity
ALF044241.AE100 mL
Catalog number ALF044241.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS108-10-1
IUPAC Name4-methylpentan-2-one
Molecular FormulaC6H12O
InChI KeyNTIZESTWPVYFNL-UHFFFAOYSA-N
SMILESCC(C)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<15 APHA
Infrared spectrumConforms
GC>=98.5 %
Water=<0.1 % (K.F.)