Magnesium oxide, 96%, heavy, Thermo Scientific Chemicals
Magnesium oxide, 96%, heavy, Thermo Scientific Chemicals
Magnesium oxide, 96%, heavy, Thermo Scientific Chemicals
Magnesium oxide, 96%, heavy, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium oxide, 96%, heavy, Thermo Scientific Chemicals
CAS: 1309-48-4 | MgO | 40.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Catalog number ALF044237.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS811-98-3
IUPAC Name(²H₃)methan(²H)ol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-MZCSYVLQSA-N
SMILES[2H]OC([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA≥99.94% Isotopic enrichment by ¹H nmr
Appearance (Color)Clear colorless
Assay from Supplier's CofA≥99.5% ¹H nmr
Water Content (Karl Fischer Titration)≤ 300ppm
Magnesium oxide is used as a laboratory reagent and an insulator in industrial cables. It acts as a reference material for calorimetry. It is widely used as a basic refractory material for crucibles, a protective coating in plasma displays, an oxide barrier in spin-tunneling devices, an optical material, a component of tubular construction in heating elements, and also in libraries for preserving books. Magnesium oxide microspheres used in enrichment method of polycyclic aromatic hydrocarbons.