Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALF044231.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS120-32-1
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
Appearance (Color)Pale cream to cream to orange or pale pink to pink
FormPowder
Melting Point (clear melt)43.0-52.0?C
Used as flavor and fragrance intermediates. It is an important raw material used intermediate in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals.