Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octakis(trimethylsiloxy)silsesquioxane, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALF044231.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialOctakis (trimethylsiloxy)silsesquioxane
CAS51777-38-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Used as flavor and fragrance intermediates. It is an important raw material used intermediate in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals.