Sodium nitrite, 99.999% (metals basis), Thermo Scientific Chemicals
Sodium nitrite, 99.999% (metals basis), Thermo Scientific Chemicals
Sodium nitrite, 99.999% (metals basis), Thermo Scientific Chemicals
Sodium nitrite, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium nitrite, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 7632-00-0 | NNaO2 | 69.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF044227.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS5431-44-7
IUPAC Namepyridine-2,6-dicarbaldehyde
Molecular FormulaC7H5NO2
InChI KeyPMWXGSWIOOVHEQ-UHFFFAOYSA-N
SMILESO=CC1=CC=CC(C=O)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point122°C to 126°C
GC>=96.0 %
Appearance (Color)White to tan
Appearance (Form)Powder
Infrared spectrumConforms
Sodium nitrite is used in many industrial applications viz. in meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, an antidote in cyanide poisoning, an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor and in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.