Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Wetting and solubilizing agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF044203.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDioctyl sulfosuccinate sodium salt
CAS577-11-7
Health Hazard 1H315-H318
Health Hazard 2GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
View more
Wetting and solubilizing agent.Dioctyl sulfosuccinate sodium salt is used as a surfactant, wetting agent and in the preparation of reverse micelles. It is utilized in electrophoresis detection of natural and synthetic estrogens. It also finds use in the drug, cosmetics, food industry and as a laxative to treat constipation. Further, it is used as a food additive, emulsifier and dispersant. It plays an important role as an excipient in the production of tablets and suspensions.