Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals
Wetting and solubilizing agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF044203.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS298-14-6
IUPAC Namepotassium hydrogen carbonate
Molecular FormulaCHKO3
InChI KeyTYJJADVDDVDEDZ-UHFFFAOYSA-M
SMILES[K+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder or crystalline powder
Titration with HCl98.5 to 101.5 %
Insoluble matter=<0.01 %
Heavy metals (as Pb)=<5 ppm
Wetting and solubilizing agent.Dioctyl sulfosuccinate sodium salt is used as a surfactant, wetting agent and in the preparation of reverse micelles. It is utilized in electrophoresis detection of natural and synthetic estrogens. It also finds use in the drug, cosmetics, food industry and as a laxative to treat constipation. Further, it is used as a food additive, emulsifier and dispersant. It plays an important role as an excipient in the production of tablets and suspensions.