Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals
Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals
Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals
Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Neodymium powder, -40 mesh, 99.8% (REO), Thermo Scientific Chemicals
CAS: 7440-00-8 | Nd | 144.24 g/mol
Have Questions?
Catalog NumberQuantity
ALF044181.065 g
Catalog number ALF044181.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS111-77-3
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4250 to 1.4270 (20°C, 589 nm)
Appearance (Form)Clear liquid
GC>=98.5 %
Color scale=<15 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only