Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Crosslinking monomer
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF044151.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS25322-68-3
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid as melt or white low-melting solid
pH4.5-7.5 (5%, demin water)
Hydroxyl Value178-197 mg KOH/g
Appearance (Color)Colorless
CommentAverage mol weight (based on OH value) 570-630
Ethylene glycol dimethacrylate is used as a functional monomer for polymers and as a cross linking agent between the molecular chains of polymers and elastomers. It is also used in free radical copolymer cross linking reactions. It acts as an intermediate in the production of hydroxyapatite and poly methyl methacrylate composites.