Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Crosslinking monomer
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALF044151.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS1116-76-3
IUPAC Nametrioctylamine
Molecular FormulaC24H51N
InChI KeyXTAZYLNFDRKIHJ-UHFFFAOYSA-N
SMILESCCCCCCCCN(CCCCCCCC)CCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4480 to 1.4500 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Ethylene glycol dimethacrylate is used as a functional monomer for polymers and as a cross linking agent between the molecular chains of polymers and elastomers. It is also used in free radical copolymer cross linking reactions. It acts as an intermediate in the production of hydroxyapatite and poly methyl methacrylate composites.