Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals
Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals
Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals
Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals
CAS: 63231-67-4 | H2O3Si | 78.098 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF044101.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS30766-11-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %