Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals
Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals
Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals
Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals
CAS: 63231-67-4 | H2O3Si | 78.098 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number ALF044101.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS13520-92-8
Molecular FormulaCl2H18O9Zr
InChI KeyFUBACIUATZGHAC-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.Cl.Cl.O=[Zr]
Molecular Weight (g/mol)324.26
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Water42 to 50 %
Titration Argentometric97.5 to 102.5 %
Solubility(10% in water) Clear to slightly hazy solution
Appearance (Form)Crystalline powder