Cobalt powder, -400 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Cobalt powder, -400 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Cobalt powder, -400 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Cobalt powder, -400 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt powder, -400 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF044085.30250 g
Catalog number ALF044085.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS71-36-3
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.2 % (Coulometric)
GC>=98.5 %
Refractive index1.3980 to 1.4000 (20°C, 589 nm)
Appearance (Form)Clear liquid