Amberlyst™ 15(H), wet, ion exchange resin, Thermo Scientific Chemicals
Amberlyst™ 15(H), wet, ion exchange resin, Thermo Scientific Chemicals
Amberlyst™ 15(H), wet, ion exchange resin, Thermo Scientific Chemicals
Amberlyst™ 15(H), wet, ion exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Amberlyst™ 15(H), wet, ion exchange resin, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF044079.30250 g
Catalog number ALF044079.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil
Thickness0.5mm (0.02in)
Total Metal Impurities0.01% max
Amberlyst™ 15(H) is a strongly acidic cation exchange resin used as a heterogeneous acid catalysis and is suitable for non-aqueous catalysis. It is used to determine the mechanism of phenol alkylation using density functional theory (DFT). It is widely utilized as a column packing material for HPLC analysis. It is employed in separation media for Thin-layer chromatography (TLC) and paper chromatography. It is used in various chemical reactions like esterification, transesterification, Michael addition, Prins cyclization, Friedel-Crafts alkylation, acylation, cleavage of epoxides and crossed-aldol condensation.