p-Toluenesulfonic acid monohydrate, ACS, 98.5+%, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS, 98.5+%, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS, 98.5+%, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS, 98.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS, 98.5+%, Thermo Scientific Chemicals
CAS: 6192-52-5 | C7H10O4S | 190.213 g/mol
Have Questions?
Catalog NumberQuantity
ALF044002.22100 g
Catalog number ALF044002.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS865-49-6
IUPAC Nametrichloro(²H)methane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-MICDWDOJSA-N
SMILES[2H]C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
D-Enrichment>=99.80 %
Water=<0.02 % (Coulometric)
TMS0.02 to 0.04 v/v %
View more
p-Toluenesulfonic acid monohydrate is used as a catalyst in the synthesis of resveratrol, in oxane derivatives as an antimalarial agent, as substituted piperidine and unsymmetrical benzyl. It acts as a catalyst in the preparation of 1,3,5-trisubstituted pyrazoles derivatives, selenated ketene dithioacetals, triazoloquinazolinone and benzimidazoquinazolinone derivatives. It also serves as an intermediate in the esterification and in reductive amination reactions.