1-Aminopyrene, 97%, Thermo Scientific Chemicals
1-Aminopyrene, 97%, Thermo Scientific Chemicals
1-Aminopyrene, 97%, Thermo Scientific Chemicals
1-Aminopyrene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Aminopyrene, 97%, Thermo Scientific Chemicals
CAS: 1606-67-3 | C16H11N | 217.27 g/mol
Have Questions?
Catalog NumberdimQuantity
ALF043968.04
Catalog number ALF043968.04
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight217.27
Unit Size
It acts as a precursor molecule for production of other fluorescent probes such as 1-Azidopyrene, APTS, and N-(1-Pyrene)iodoacetamide.