N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, Thermo Scientific Chemicals
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, Thermo Scientific Chemicals
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, Thermo Scientific Chemicals
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% TMCS, Thermo Scientific Chemicals
CAS: 25561-30-2 | C8H18F3NOSi2 | 257.403 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALF043939.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN,O-Bis (trimethylsilyl)trifluoroacetamide
Name NoteWith 1% TMCS
CAS25561-30-2
Health Hazard 1H226-H314
Health Hazard 2GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
View more
N,O-Bis(trimethylsilyl)trifluoroacetamide is used as a derivatization reagent for gas chromatography and mass-spectrometry analysis of phenolic acids in fruits. It is also used as a reactant in the preparation of pyrimidinone ribosides and analogs which shows anti-tumor properties. It plays as an important role as a reagent for the regioselective desulfation of polysaccharide sulfates. Furthermore, it is used to prepare trimethylsilyl ethers of carbohydrates and alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only