Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium cobalt(II) sulfate hexahydrate, 99%, Thermo Scientific Chemicals
As spectroscopic standard, cobalt plating, catalysis, ceramics
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF043902.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS90-04-0
IUPAC Name2-methoxyaniline
Molecular FormulaC7H9NO
InChI KeyVMPITZXILSNTON-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.0 %
Appearance (Color)Clear colorless to yellow-brown
Infrared spectrumConforms
Refractive index1.5730 to 1.5755 (20°C, 589 nm)
Ammonium cobalt(II) sulfate hexahydrate is uses as spectroscopic standard, cobalt plating, catalysis and ceramics. It is also used in pharmaceutical intermediates.