Aluminum oxide, Alpha-phase, Catalyst support, Low surface area, Trimodal, Thermo Scientific Chemicals
Aluminum oxide, Alpha-phase, Catalyst support, Low surface area, Trimodal, Thermo Scientific Chemicals
Aluminum oxide, Alpha-phase, Catalyst support, Low surface area, Trimodal, Thermo Scientific Chemicals
Aluminum oxide, Alpha-phase, Catalyst support, Low surface area, Trimodal, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminum oxide, Alpha-phase, Catalyst support, Low surface area, Trimodal, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF043833.A11 kg
Catalog number ALF043833.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS261952-04-9
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4605-1.4655 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%