Magnesium silicate, 99% (metals basis), Thermo Scientific Chemicals
Magnesium silicate, 99% (metals basis), Thermo Scientific Chemicals
Magnesium silicate, 99% (metals basis), Thermo Scientific Chemicals
Magnesium silicate, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium silicate, 99% (metals basis), Thermo Scientific Chemicals
CAS: 1343-88-0 | MgO3Si | 100.39 g/mol
Have Questions?
Catalog NumberQuantity
ALF043807.1850 g
Catalog number ALF043807.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMagnesium silicate
Melting Point1910°C
CAS1343-88-0
ColorWhite
Health Hazard 1H332: Harmful if inhaled.
View more
Magnesium silicate is employed for polyether adsorbent, which has applications of adsorption and decoloring, adsorbent in gas chromatography, in vitamin analysis, antibiotic processing, ant caking agent, used in cosmetics as talcum & baby powder, general modified plastics, mid-range insulation materials, EVA shoes material.