Sodium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals
Sodium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals
Sodium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals
Sodium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, 25% w/v aq. soln., Thermo Scientific Chemicals
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Catalog NumberQuantity
ALF043771.K21 L
Catalog number ALF043771.K2
Price (MYR)
320.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
320.00
Add to cart
Specifications
Chemical Name or MaterialSodium hydroxide
ColorColorless
CAS1310-73-2
Health Hazard 1H290-H314-H335
Health Hazard 2In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).