Polystyrene latex microsphere, 1 micron, dry form
Polystyrene latex microsphere, 1 micron, dry form
Polystyrene latex microsphere, 1 micron, dry form
Polystyrene latex microsphere, 1 micron, dry form
Thermo Scientific Chemicals
Polystyrene latex microsphere, 1 micron, dry form
CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
ALF043711.ME500 mg
Catalog number ALF043711.ME
Price (MYR)
4,067.00
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (MYR)
4,067.00
Add to cart
Specifications
Chemical Name or MaterialPolystyrene latex microsphere
Name NoteDry form
CAS9003-53-6
Physical FormMicrosphere
Recommended StorageAmbient temperatures
View more
Polystyrene is used in adhesive, plastics, coatings, for electrical applications. In laboratories containers such as tubes, petridishes were made in polystyrene, used for the building construction panel packing, insulators, for injection molding and casting, as sheets, plates, rods, rigid forms and beads..