N,N'-Methylenebisacrylamide, 99+%
N,N'-Methylenebisacrylamide, 99+%
N,N'-Methylenebisacrylamide, 99+%
N,N'-Methylenebisacrylamide, 99+%
Thermo Scientific Chemicals
N,N'-Methylenebisacrylamide, 99+%
CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF043701.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS344-03-6
IUPAC Name1,4-dibromo-2,3,5,6-tetrafluorobenzene
Molecular FormulaC6Br2F4
InChI KeyQFTZULJNRAHOIY-UHFFFAOYSA-N
SMILESFC1=C(F)C(Br)=C(F)C(F)=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder or chunks
Melting Point (clear melt)75.0-82.0?C
Bisacrylamide is used in biochemistry as a preparatory agent for polyacrylamide gel for electrophoretic separation of nucleic acids and proteins.