N,N'-Methylenebisacrylamide, 99+%
N,N'-Methylenebisacrylamide, 99+%
N,N'-Methylenebisacrylamide, 99+%
N,N'-Methylenebisacrylamide, 99+%
Thermo Scientific Chemicals
N,N'-Methylenebisacrylamide, 99+%
CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALF043701.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN,N'-Methylenebisacrylamide
CAS110-26-9
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
Bisacrylamide is used in biochemistry as a preparatory agent for polyacrylamide gel for electrophoretic separation of nucleic acids and proteins.