Deuterium oxide, 99.95+% (Isotopic), Thermo Scientific Chemicals
Deuterium oxide, 99.95+% (Isotopic), Thermo Scientific Chemicals
Deuterium oxide, 99.95+% (Isotopic), Thermo Scientific Chemicals
Deuterium oxide, 99.95+% (Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Deuterium oxide, 99.95+% (Isotopic), Thermo Scientific Chemicals
CAS: 7789-20-0 | H2O | 20.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 Pk.
10 Pk.
Catalog number ALF043700.SA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 Pk.
Request bulk or custom format
Chemical Identifiers
CAS550-99-2
IUPAC Namehydrogen 2-[(naphthalen-1-yl)methyl]-4,5-dihydro-1H-imidazole chloride
Molecular FormulaC14H15ClN2
InChI KeyDJDFFEBSKJCGHC-UHFFFAOYSA-N
SMILES[H+].[Cl-].C(C1=NCCN1)C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Assay (HPLC)≥98.5%
Identification (FTIR)Conforms
Deuterium oxide is used in nuclear magnetic resonance spectroscopy (NMR). It is also useful in the identification of labile hydrogens. As a source of deuterium, it is utilized for preparing specifically labeled isotopologs of organic compounds. It is often used instead of water for analyzing proteins in solution by using fourier transform infrared spectroscopy (FTIR). It finds application in certain types of nuclear reactors and in tritium production.