1-Hydroxyethylidenebis(phosphonic acid), 96%, Thermo Scientific Chemicals
1-Hydroxyethylidenebis(phosphonic acid), 96%, Thermo Scientific Chemicals
1-Hydroxyethylidenebis(phosphonic acid), 96%, Thermo Scientific Chemicals
1-Hydroxyethylidenebis(phosphonic acid), 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Hydroxyethylidenebis(phosphonic acid), 96%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF043502.1850 g
Catalog number ALF043502.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS549-18-8
IUPAC Namehydrogen dimethyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}propyl)amine chloride
Molecular FormulaC20H24ClN
InChI KeyKFYRPLNVJVHZGT-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CCC=C1C2=CC=CC=C2CCC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay (HPLC)98.0% min.
Loss on Drying6.0% max.
Melting Point195-200?C
FormPowder
1-Hydroxyethylidenebis(phosphonic acid) acts as human protein tyrosine phosphatase inhibitor. It is also useful for photographic applications. It finds application to strengthen bone, osteoporosis treatment and to treat Paget's disease of bone. In addition, it is used in corrosion inhibition in circulating cool water systems, oil fields and low-pressure boilers with electric power, the chemical industry, metallurgy and fertilizers. It is also utilized for formulating reverse osmosis and thermal desalination anti-scaling agents. Furthermore, it is used in detergents, water treatment, cosmetics and pharmaceuticals.