2-Propanethiol, 98%, Thermo Scientific Chemicals
2-Propanethiol, 98%, Thermo Scientific Chemicals
2-Propanethiol, 98%, Thermo Scientific Chemicals
2-Propanethiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Propanethiol, 98%, Thermo Scientific Chemicals
CAS: 75-33-2 | C3H8S | 76.157 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number ALF043422.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS1809-19-4
IUPAC Namedibutoxy(oxo)-λ⁵-phosphanylium
Molecular FormulaC8H18O3P
InChI KeyOSPSWZSRKYCQPF-UHFFFAOYSA-N
SMILESCCCCO[P+](=O)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay14.5 to 16 % (P)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4225 to 1.4255 (20°C, 589 nm)
2-Propanethiol Reacts with 3-chloro-propene to get 3-isopropylsulfanyl-propene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only