1-Propanethiol, 98%, Thermo Scientific Chemicals
1-Propanethiol, 98%, Thermo Scientific Chemicals
1-Propanethiol, 98%, Thermo Scientific Chemicals
1-Propanethiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Propanethiol, 98%, Thermo Scientific Chemicals
CAS: 107-03-9 | C3H8S | 76.157 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number ALF043420.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or Material1-Propanethiol
CAS107-03-9
Health Hazard 1H225-H302-H315-H318-H335
Health Hazard 2GHS H Statement
H225-H318-H302-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
View more
1-Propanethiol, is used widely in research areas. This compound has a wide range of applications including cell biology, materials science, nanoscience, self-assembled monolayers (SAMs), nanoelectronics, and contact printing. It is also used as a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only