Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%, Thermo Scientific Chemicals
Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%, Thermo Scientific Chemicals
Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%, Thermo Scientific Chemicals
Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%, Thermo Scientific Chemicals
Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%, Thermo Scientific Chemicals
CAS: 32740-79-7 | O2Ru | 133.07 g/mol
Have Questions?
Catalog NumberQuantity
ALF043403.065 g
Catalog number ALF043403.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Colorless to pale yellow
Color (APHA)<30
Assay (GC)≥99.5% (Corrected for water)
CommentInert gas used can be either Argon or Nitrogen
View more
Chemical catalyst, important material for producing resistance and capacitor.