Methyl levulinate, 99+%, Thermo Scientific Chemicals
Methyl levulinate, 99+%, Thermo Scientific Chemicals
Methyl levulinate, 99+%, Thermo Scientific Chemicals
Methyl levulinate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl levulinate, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF043397.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS111-84-2
IUPAC Namenonane
Molecular FormulaC9H20
InChI KeyBKIMMITUMNQMOS-UHFFFAOYSA-N
SMILESCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.4040-1.4070 @ 20?C
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Methyl levulinate is mainly used as raw material on perfume.