Iridium(IV) oxide, Premion™, 99.99% (metals basis), Ir 84.5% min, Thermo Scientific Chemicals
Iridium(IV) oxide, Premion™, 99.99% (metals basis), Ir 84.5% min, Thermo Scientific Chemicals
Iridium(IV) oxide, Premion™, 99.99% (metals basis), Ir 84.5% min, Thermo Scientific Chemicals
Iridium(IV) oxide, Premion™, 99.99% (metals basis), Ir 84.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iridium(IV) oxide, Premion™, 99.99% (metals basis), Ir 84.5% min, Thermo Scientific Chemicals
CAS: 12030-49-8 | CH4 | 16.04 g/mol
Have Questions?
Catalog NumberQuantity
ALF043396.031 g
Catalog number ALF043396.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS111-88-6
IUPAC Nameoctane-1-thiol
Molecular FormulaC8H18S
InChI KeyKZCOBXFFBQJQHH-UHFFFAOYSA-N
SMILESCCCCCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless
GC>=96.0 %
Refractive index1.4525 to 1.4545 (20°C, 589 nm)
Iridium(IV) oxide is used in the coating of anode-electrodes for industrial electrolysis in association with other rare oxides. It is also used in microelectrodes for electrophysiology research.