Paraformaldehyde, 16% w/v aq. soln., methanol free
Paraformaldehyde, 16% w/v aq. soln., methanol free
Paraformaldehyde, 16% w/v aq. soln., methanol free
Paraformaldehyde, 16% w/v aq. soln., methanol free
Thermo Scientific Chemicals
Paraformaldehyde, 16% w/v aq. soln., methanol free
Paraformaldehyde, CH2O, CAS Number-30525-89-4, methylene oxide, oxomethane, paraform, oxymethylene, formalin, formol, methyl aldehyde, paraformaldehyde, formic aldehyde, methanal, 10 x 10mL, CHEBI:16842, Hazard Class: 9; Packaging Group: III, 200-001-8, H312+H332-H315-H317-H319-H335-H341-H350
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 x 10 mL
10 x 10 mL
Catalog number ALF043368.9L
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 x 10 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialParaformaldehyde
Name NoteMethanol free
Health Hazard 1H312+H332-H315-H317-H319-H335-H341-H350
Health Hazard 2GHS H Statement
H351-H315-H319-H317
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P261-P264b-P271-P272-P280-P281-P301+P330+P331-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
View more
Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only