Palladium granules, 99.95% (metals basis), Thermo Scientific Chemicals
Palladium granules, 99.95% (metals basis), Thermo Scientific Chemicals
Palladium granules, 99.95% (metals basis), Thermo Scientific Chemicals
Palladium granules, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Palladium granules, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALF043353.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS92-86-4
IUPAC Name4,4'-dibromo-1,1'-biphenyl
Molecular FormulaC12H8Br2
InChI KeyHQJQYILBCQPYBI-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder or crystals
Melting point165°C to 169°C