Hydrochloric acid, 5% v/v aq. soln., Thermo Scientific Chemicals
Hydrochloric acid, 5% v/v aq. soln., Thermo Scientific Chemicals
Hydrochloric acid, 5% v/v aq. soln., Thermo Scientific Chemicals
Hydrochloric acid, 5% v/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, 5% v/v aq. soln., Thermo Scientific Chemicals
CAS: 7647-10-1 | Cl2Pd | 177.32 g/mol
Have Questions?
Catalog NumberQuantity
ALF043335.K21 L
Catalog number ALF043335.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS110-42-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
FormLiquid
Refractive Index1.4240-1.4270 @ 20°C