Silicon carbide, 99% (metals basis), Thermo Scientific Chemicals
Silicon carbide, 99% (metals basis), Thermo Scientific Chemicals
Silicon carbide, 99% (metals basis), Thermo Scientific Chemicals
Silicon carbide, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Silicon carbide, 99% (metals basis), Thermo Scientific Chemicals
CAS: 409-21-2 | CSi | 40.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number ALF043332.A3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialSilicon carbide
Melting Point2700°C
CAS409-21-2
ColorBlack
Health Hazard 1Warning
View more
Silicon carbide powder is used as an abrasive for such as grinding wheels, whetstone, grinding wheel and sand tiles. Silicon carbide is used to produce epitaxial grapheme by graphitization at high temperatures. It is also acts asthe metallurgical deoxidizer material. Four main application areas of SiC: functional ceramics, advanced refractory, abrasives and metallurgical materials. High purity single crystal, is used to manufacture semiconductors, manufacture of silicon carbide fibers. Silicon carbide is very hard, with excellent thermal conductivity, as a semiconductor and high temperature resistant to oxidation.