Brilliant Blue G, ultrapure
Brilliant Blue G, ultrapure
Brilliant Blue G, ultrapure
Brilliant Blue G, ultrapure
Thermo Scientific Chemicals
Brilliant Blue G, ultrapure
Protein staining following gel electrophoresis
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number ALF043318.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS1476-53-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to yellow
Residue on ignition10.5-12.0%
Impurity content≤10.0% (HPLC)
Loss on Drying≤6%
Potency≥850?g/mg (dried basis)
View more
Brilliant Blue G is used in analytical biochemistry dye and as a P2X7 antagonist. It is also used in protein staining following gel electrophoresis. It is used to prepare the protein reagent for the determination of protein content of the collagenase enzyme isolated from fish waste. It may be employed as stain for the internal limiting membrane (ILM) for the macular hole (MH) and epiretinal membrane (ERM) surgery. It is useful for SDS gels as it readily stains proteins with minimal background color. Protein bands can be visualized during staining. It has been used in the Bradford dye-binding protein assay. A mechanism for dye binding to protein has been proposed, based on measuring Coomassie Brilliant Blue G-250 (CBBG) absorbance spectra during titration of the dye reagent in the absence of protein and its response to different polyamino acids. Brilliant Blue G has been used for determining critical micelle concentration of detergents.