Cobalt wire, 2.0mm (0.08in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Cobalt wire, 2.0mm (0.08in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Cobalt wire, 2.0mm (0.08in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Cobalt wire, 2.0mm (0.08in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt wire, 2.0mm (0.08in) dia, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7440-48-4 | Co | 58.93 g/mol
Have Questions?
Catalog NumberQuantity
ALF043290.BQ5 cm
Catalog number ALF043290.BQ
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 cm
Request bulk or custom format
Chemical Identifiers
CAS40101-17-5
IUPAC Namebis(3-methoxyphenyl)ethane-1,2-dione
Molecular FormulaC16H14O4
InChI KeyPJGXOGKIVAJFTE-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1)C(=O)C(=O)C1=CC(OC)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or granules
CommentSpecification differs for U.S. and non-U.S. material where indicated
Appearance (Color)Pale yellow to yellow
Assay (GC)≥99.0%
Melting Point (clear melt)79.0-85.0°C